Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olfr1289 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146404.1 | NP_666516.1 | 1 | 750 | ||
GenBank mRNA | BC120736.1 | - | 1 | 832 | 40129659 | |
BC137761.1 | - | 1 | 833 | 8860780 |