Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Olfr1370 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146535.1 | NP_666746.1 | 1 | 737 | ||
GenBank mRNA | BC110468.1 | - | 1 | 737 | 40043199 | |
BC110469.1 | - | 1 | 736 |