Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olfr267 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146920.2 | NP_667131.2 | 1 | 123 | ||
GenBank mRNA | BC127983.1 | - | 1 | 126 | 40135361 |