Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olfr1494 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146990.1 | NP_667201.1 | 1 | 684 | ||
GenBank mRNA | BC104099.1 | - | 1 | 684 | 40043096 | |
BC104100.1 | - | 1 | 684 | 40043097 | ||
BC106804.1 | - | 1 | 684 | 40042568 |