Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clcn6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011929.2 | NP_036059.1 | 12 | 996 | ||
GenBank mRNA | AK045787.1 | - | 7 | 583 | ||
AK145053.1 | - | 12 | 1041 | |||
AK165362.1 | - | 12 | 1045 |