Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mapk3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011952.2 | NP_036082.1 | 3 | 533 | ||
GenBank mRNA | AK155287.1 | - | 2 | 475 | ||
BC013754.1 | - | 3 | 366 | |||
BC029712.1 | - | 3 | 527 | 5041517 | ||
BQ929318.1 | - | 3 | 533 | |||
Z14249.1 | - | 3 | 504 |