Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp260 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011981.4 | NP_036111.2 | 3 | 799 | ||
GenBank mRNA | AK088177.1 | - | 1 | 321 | ||
BC082570.1 | - | 1 | 125 | 30607787 | ||
BC085180.1 | - | 3 | 799 | 30639007 | ||
D45210.1 | - | 2 | 611 |