Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cops5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277101.1 | NP_001264030.1 | 2 | 433 | ||
NM_013715.2 | NP_038743.1 | 2 | 682 | |||
GenBank mRNA | AF065386.1 | - | 2 | 435 | ||
AF068223.1 | - | 2 | 450 | |||
AK002761.1 | - | 2 | 464 | |||
AK005728.1 | - | 2 | 469 | |||
AK012499.1 | - | 2 | 469 | |||
AK133073.1 | - | 2 | 462 | |||
AK146271.1 | - | 2 | 469 | |||
AK151492.1 | - | 2 | 468 | |||
BC046753.1 | - | 2 | 428 | 5693100 | ||
CX240457.1 | - | 2 | 650 | |||
DV056728.1 | - | 2 | 489 | |||
U70736.1 | - | 2 | 458 |