Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cops5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277101.1 | NP_001264030.1 | 3 | 546 | ||
NM_013715.2 | NP_038743.1 | 3 | 795 | |||
GenBank mRNA | AF065386.1 | - | 3 | 548 | ||
AF068223.1 | - | 3 | 563 | |||
AK002761.1 | - | 3 | 577 | |||
AK005728.1 | - | 3 | 582 | |||
AK012499.1 | - | 3 | 582 | |||
AK133073.1 | - | 3 | 575 | |||
AK146271.1 | - | 3 | 582 | |||
AK151492.1 | - | 3 | 581 | |||
BC046753.1 | - | 3 | 541 | 5693100 | ||
DV056728.1 | - | 3 | 602 | |||
U70736.1 | - | 3 | 571 |