Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bahcc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198423.3 | NP_940815.3 | 21, 22 | 6214 | ||
GenBank mRNA | AB257855.1 | - | 20, 21 | 6216 | ||
AK147250.1 | - | 21, 22 | 6214 | |||
AK147400.1 | - | 13, 14 | 3122 | |||
AK164229.1 | - | 4, 5 | 455 | |||
BC060615.1 | - | 4, 5 | 486 | |||
BC072602.1 | - | 10, 11 | 1652 | 6830770 |