Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arhgef40 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145921.1 | NP_001139393.1 | 3 | 1489 | ||
NM_001145922.1 | NP_001139394.1 | 3 | 1489 | |||
NM_198249.4 | NP_937892.2 | 3 | 1489 | |||
GenBank mRNA | AK053592.1 | - | 3 | 1421 | ||
AK131136.1 | - | 3 | 1458 | |||
AK134295.1 | - | 3 | 1478 | |||
AK143527.1 | - | 3 | 1464 | |||
AK147761.1 | - | 3 | 1425 | |||
AK155659.1 | - | 2 | 2508 | |||
AK163945.1 | - | 3 | 1490 | |||
BC059018.1 | - | 3 | 1391 | 6848691 |