Gene Symbol: Rnf31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_194346.2 | NP_919327.2 | 19 | 3030 | ||
GenBank mRNA | AK131160.1 | - | 12 | 4589 | ||
AK156320.1 | - | 19 | 3113 | |||
AY283085.1 | - | 19 | 2939 | |||
BC057595.1 | - | 19 | 3030 | 6811051 |