Gene Symbol: Rnf31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_194346.2 | NP_919327.2 | 21 | 3276 | ||
GenBank mRNA | AK131160.1 | - | 14 | 4835 | ||
AK156320.1 | - | 21 | 3359 | |||
AY283085.1 | - | 21 | 3185 | |||
BC057595.1 | - | 21 | 3276 | 6811051 |