Search Thermo Fisher Scientific
Gene Symbol: B3galt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020025.4 | NP_064409.3 | 2 | 1500 | ||
GenBank mRNA | AK034371.1 | - | 2 | 1456 | ||
AK036141.1 | - | 2 | 1502 | |||
AK083168.1 | - | 2 | 1455 | |||
BC046322.1 | - | 2 | 1336 | 6392262 |