Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: B3galnt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020026.4 | NP_064410.1 | 3 | 1068 | ||
GenBank mRNA | AK003837.1 | - | 1 | 582 | ||
AK088407.1 | - | 3 | 971 | |||
BC003835.1 | - | 2 | 874 | 3486016 |