Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mprip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012027.2 | NP_036157.2 | 14 | 2015 | ||
NM_201245.3 | NP_957697.1 | 14 | 2015 | |||
GenBank mRNA | AB093269.1 | - | 1 | 36 | ||
AK035435.1 | - | 8 | 1305 | |||
AK044512.1 | - | 13 | 1657 | |||
AK154807.1 | - | 15 | 2022 | |||
AK155630.1 | - | 14 | 1796 | |||
AK158296.1 | - | 14 | 2004 | |||
AK162530.1 | - | 12 | 1361 | |||
AK170849.1 | - | 14 | 1999 | |||
AK171089.1 | - | 14 | 2004 | |||
BC039932.1 | - | 13 | 1539 | |||
BC049803.1 | - | 14 | 1541 | |||
BC063067.1 | - | 14 | 1978 | 5721396 | ||
U73200.1 | - | 14 | 1875 |