Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mprip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012027.2 | NP_036157.2 | 16 | 2534 | ||
NM_201245.3 | NP_957697.1 | 16 | 2534 | |||
GenBank mRNA | AB093269.1 | - | 4 | 4359 | ||
AK035435.1 | - | 10 | 1824 | |||
AK154807.1 | - | 17 | 2541 | |||
AK155630.1 | - | 16 | 2315 | |||
AK158296.1 | - | 16 | 2523 | |||
AK162530.1 | - | 14 | 1880 | |||
AK165710.1 | - | 2 | 693 | |||
AK170849.1 | - | 16 | 2518 | |||
AK171089.1 | - | 16 | 2523 | |||
BC027417.1 | - | 2 | 623 | |||
BC039932.1 | - | 15 | 2058 | |||
BC049803.1 | - | 16 | 2060 | |||
BC063067.1 | - | 16 | 2497 | 5721396 | ||
U73200.1 | - | 16 | 2394 |