Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Mprip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012027.2 | NP_036157.2 | 6 | 921 | ||
NM_201245.3 | NP_957697.1 | 6 | 921 | |||
GenBank mRNA | AK044512.1 | - | 6 | 671 | ||
AK154807.1 | - | 6 | 928 | |||
AK155630.1 | - | 6 | 702 | |||
AK158296.1 | - | 6 | 910 | |||
AK162530.1 | - | 3 | 252 | |||
AK170849.1 | - | 6 | 905 | |||
AK171089.1 | - | 6 | 910 | |||
BC039932.1 | - | 5 | 445 | |||
BC063067.1 | - | 6 | 884 | 5721396 | ||
U73200.1 | - | 6 | 781 |