Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tspan12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173007.3 | NP_766595.1 | 7 | 709 | ||
GenBank mRNA | AK033554.1 | - | 6 | 544 | ||
AK050737.1 | - | 7 | 710 | |||
BC065152.1 | - | 6 | 520 | 6808971 | ||
BC068240.1 | - | 6 | 571 | 3674585 |