Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Shroom3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077595.2 | NP_001071063.1 | 10 | 5789 | ||
NM_001077596.2 | NP_001071064.1 | 10 | 5837 | |||
NM_015756.2 | NP_056571.2 | 11 | 6161 | |||
GenBank mRNA | AF199421.1 | - | 11 | 6179 | ||
AF199422.1 | - | 10 | 5747 | |||
AK003320.1 | - | 2 | 948 | |||
AK049347.1 | - | 7 | 3695 | |||
AK129371.1 | - | 7 | 5167 | |||
AK147493.1 | - | 10 | 5772 | |||
AK164918.1 | - | 10 | 5748 | |||
BC003909.1 | - | 5 | 1433 | |||
BC145113.1 | - | 12 | 5578 | 9053661 |