Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pcdh19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105245.1 | NP_001098715.1 | 1 | 2411 | ||
NM_001105246.1 | NP_001098716.1 | 1 | 2411 | |||
GenBank mRNA | AK046726.1 | - | 1 | 2261 | ||
AK122492.1 | - | 1 | 154 | |||
BC118529.1 | - | 2 | 600 | 40098492 |