Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Usp39 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138592.4 | NP_613058.1 | 11 | 1571 | ||
GenBank mRNA | AK010443.1 | - | 11 | 1581 | ||
AK018595.1 | - | 11 | 1572 | |||
AK033105.1 | - | 11 | 1568 | |||
AK045062.1 | - | 11 | 1568 | |||
AK088354.1 | - | 11 | 1583 | |||
AK165625.1 | - | 11 | 1552 | |||
AK167668.1 | - | 11 | 1559 | |||
BC026983.1 | - | 11 | 1546 | 5365862 |