Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slco1b2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020495.1 | NP_065241.1 | 15 | 2020 | ||
GenBank mRNA | AB031959.1 | - | 15 | 2020 | ||
AF250912.1 | - | 7 | 1148 |