Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slco1b2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020495.1 | NP_065241.1 | 5, 6 | 555 | ||
GenBank mRNA | AB031959.1 | - | 5, 6 | 555 | ||
AK144557.1 | - | 1, 2 | 328 |