Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Flnb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081427.1 | NP_001074896.1 | 29 | 5180 | ||
NM_134080.1 | NP_598841.1 | 29 | 5180 | |||
GenBank mRNA | CF166040.1 | - | 3 | 333 | ||
CF536812.1 | - | 2 | 187 | |||
CF736029.1 | - | 2 | 191 |