Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Mga | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164274.1 | NP_001157746.1 | 4 | 2230 | ||
NM_013720.2 | NP_038748.2 | 4 | 2230 | |||
GenBank mRNA | AF205935.1 | - | 3 | 2044 | ||
AK032570.1 | - | 4 | 2211 | |||
AK046252.1 | - | 4 | 2231 | |||
AK172962.1 | - | 4 | 2153 |