Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bcar3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013867.2 | NP_038895.1 | 9, 10 | 2372 | ||
GenBank mRNA | AF179566.1 | - | 9, 10 | 2320 | ||
AK143894.1 | - | 9, 10 | 2160 | |||
AK158241.1 | - | 9, 10 | 2373 | |||
BC023930.1 | - | 9, 10 | 2361 | 5324365 |