Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pdlim4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019417.3 | NP_062290.2 | 7 | 949 | ||
GenBank mRNA | AB221672.1 | - | 7 | 864 | ||
AI385555.1 | - | 1 | 237 | |||
BC030068.1 | - | 7 | 878 | 5144166 | ||
Y08361.1 | - | 7 | 900 |