Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc22a4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019687.3 | NP_062661.1 | 5 | 1048 | ||
GenBank mRNA | AB016257.1 | - | 5 | 1055 | ||
AK143595.1 | - | 4 | 958 | |||
BC010590.1 | - | 5 | 1048 | 3670138 |