Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adat1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013925.4 | NP_038953.1 | 2 | 193 | ||
GenBank mRNA | AF192375.1 | - | 1 | 32 | ||
AK035614.1 | - | 2 | 181 | |||
AK039699.1 | - | 2 | 163 | |||
AK082399.1 | - | 2 | 194 | |||
AK161044.1 | - | 2 | 163 | |||
BC019976.1 | - | 2 | 139 | 4458235 |