Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dpep2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_176913.4 | NP_795887.2 | 1 | 225 | ||
GenBank mRNA | AF488552.1 | - | 1 | 225 |