Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Efcab5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_176965.3 | NP_795939.2 | 18 | 4216 | ||
GenBank mRNA | AK029790.1 | - | 16 | 3804 | ||
AK163359.1 | - | 2 | 411 | |||
BC127154.1 | - | 12 | 3022 |