Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Scai | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178778.3 | NP_848893.2 | 14, 15 | 1404 | ||
GenBank mRNA | AK044772.1 | - | 14, 15 | 1400 | ||
AK082408.1 | - | 15, 16 | 1506 | |||
BC132485.1 | - | 14, 15 | 1379 | 40130762 | ||
BC144986.1 | - | 15, 16 | 1480 | |||
BC144987.1 | - | 14, 15 | 1455 | |||
BC144989.1 | - | 14, 15 | 1378 |