Search Thermo Fisher Scientific
Gene Symbol: Ccdc158 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177230.3 | NP_796204.1 | 10 | 1289 | ||
GenBank mRNA | AK029997.1 | - | 10 | 1289 | ||
BC137976.1 | - | 10 | 1258 | 8860995 | ||
BC145072.1 | - | 10 | 1259 |