Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mtap7d3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177293.3 | NP_796267.2 | 10 | 1687 | ||
GenBank mRNA | AK030211.1 | - | 9 | 1604 | ||
AK048385.1 | - | 4 | 697 | |||
BC096764.1 | - | 10 | 1689 | |||
BC118963.2 | - | 9 | 1604 | 40095274 |