Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mtap7d3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177293.3 | NP_796267.2 | 7 | 1270 | ||
GenBank mRNA | AK030211.1 | - | 6 | 1187 | ||
AK048385.1 | - | 1 | 280 | |||
AK049164.1 | - | 7 | 1170 | |||
BC096764.1 | - | 7 | 1272 | |||
BC118963.2 | - | 6 | 1187 | 40095274 |