Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyp4f39 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177307.3 | NP_796281.1 | 11, 12 | 1455 | ||
GenBank mRNA | AK028950.1 | - | 11, 12 | 1456 | ||
AK037192.1 | - | 11, 12 | 1456 | |||
BC145755.1 | - | 11, 12 | 1452 | 40131058 |