Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ubash3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177823.4 | NP_808491.2 | 12, 13 | 1687 | ||
GenBank mRNA | AK040572.1 | - | 12, 13 | 1648 | ||
AK041690.1 | - | 12, 13 | 1654 | |||
AK049106.1 | - | 11, 12 | 1587 |