Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dixdc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178118.2 | NP_835219.1 | 5 | 821 | ||
GenBank mRNA | AK164424.1 | - | 5 | 744 | ||
AY549883.1 | - | 5 | 679 | |||
AY549884.1 | - | 5 | 682 | |||
BC048182.1 | - | 5 | 812 | 6826299 | ||
BC063085.1 | - | 5 | 812 | 6826299 |