Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Cndp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177450.4 | NP_803233.2 | 3 | 406 | ||
GenBank mRNA | AK085308.1 | - | 3 | 396 | ||
AK165500.1 | - | 3 | 257 | |||
AK165706.1 | - | 3 | 358 | |||
BC043305.1 | - | 3 | 335 | 5040944 |