Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pramel7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178250.2 | NP_839981.1 | 3 | 1141 | ||
GenBank mRNA | AF490343.1 | - | 3 | 1141 | ||
BC052825.1 | - | 3 | 1152 | 30030731 |