Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem232 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008973.2 | NP_001008973.2 | 11 | 1272 | ||
NM_001289485.1 | NP_001276414.1 | 12 | 1357 | |||
NM_001289486.1 | NP_001276415.1 | 11 | 1269 | |||
GenBank mRNA | AF338468.1 | - | 9 | 1036 | ||
AF338469.1 | - | 12 | 1351 | |||
AK053319.1 | - | 5 | 606 | |||
AK135727.1 | - | 3 | 263 | |||
BC050244.1 | - | 9 | 1057 | |||
BC150885.1 | - | 11 | 1234 | 9087796 | ||
BC172032.1 | - | 11 | 1231 | 9054318 | ||
DV662151.1 | - | 3 | 232 |