Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nsl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198654.3 | NP_941056.3 | 2 | 312 | ||
GenBank mRNA | AK029421.1 | - | 2 | 310 | ||
BC029086.1 | - | 2 | 302 | 3156934 |