Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Nr1h5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198658.2 | NP_941060.2 | 4, 5 | 671 | ||
NM_201619.2 | NP_963913.2 | 4, 5 | 671 | |||
NM_201622.2 | NP_963916.2 | 4, 5 | 497 | |||
GenBank mRNA | AY094586.1 | - | 4, 5 | 671 | ||
AY094587.1 | - | 3, 4 | 566 | |||
AY094588.1 | - | 3, 4 | 566 | |||
AY094589.1 | - | 3, 4 | 566 | |||
AY094590.1 | - | 3, 4 | 566 | |||
AY094591.1 | - | 3, 4 | 392 | |||
BC119524.1 | - | 3, 4 | 566 | 40056073 | ||
BC119525.1 | - | 3, 4 | 566 | 40056074 |