Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Taok2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163774.1 | NP_001157246.1 | 8 | 1699 | ||
NM_001163775.1 | NP_001157247.1 | 8 | 1699 | |||
GenBank mRNA | AK031190.1 | - | 8 | 1435 | ||
AK129235.1 | - | 8 | 806 | |||
BC075676.1 | - | 8 | 1262 | |||
BC077714.1 | - | 8 | 1148 | |||
BC085152.1 | - | 8 | 1402 |