Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cep44 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009951.1 | NP_001009951.1 | 11 | 1208 | ||
GenBank mRNA | AK220415.1 | - | 10 | 1164 | ||
BC088983.1 | - | 11 | 1208 | 5700628 |