Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gm1110 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001281475.1 | NP_001268404.1 | 5, 6 | 549 | ||
GenBank mRNA | DQ664197.1 | - | 5, 6 | 549 |