Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tas2r121 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207024.1 | NP_996907.1 | 1 | 381 | ||
GenBank mRNA | BC104410.1 | - | 1 | 381 | 40050579 | |
BC104411.1 | - | 1 | 381 | 40050580 |