Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zhx2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199449.2 | NP_955520.1 | 3 | 2586 | ||
GenBank mRNA | AB099526.1 | - | 1 | 1969 | ||
AK031782.1 | - | 3 | 2483 | |||
AK157631.1 | - | 3 | 2573 | |||
BC059178.1 | - | 3 | 2340 | 6400319 |