Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ugt1a7c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_201642.4 | NP_964004.1 | 1 | 879 | ||
GenBank mRNA | AK128918.1 | - | 1 | 805 | ||
AK144599.1 | - | 1 | 771 | |||
AK153157.1 | - | 1 | 769 | |||
AK154973.1 | - | 1 | 802 | |||
AK159496.1 | - | 1 | 817 | |||
AK162317.1 | - | 1 | 803 | |||
AK165465.1 | - | 1 | 803 | |||
AY227199.1 | - | 1 | 783 | |||
BC141205.1 | - | 1 | 788 | 9055817 | ||
BC141206.1 | - | 1 | 788 | 9055818 |